Learn More
Med Vet International Defensor 1 , Rabies Vaccine, Killed Virus, 50 X 1mL
Catalog No. 502828392
Encompass_Preferred
VET LICENSE NEEDS TO BE PROVIDED IN 3-4 BUSINESS DAYS OR ORDER WILL BE CANCELLED
Rabies Vaccine, Killed Virus, dogs and cats 3 months and older. DOI is 1 year. Preservative: Gentamicin.
Catalog No. 50-282-8392 Supplier Med Vet International Supplier No. RXV1RABDEF50D
VET LICENSE NEEDS TO BE PROVIDED IN 3-4 BUSINESS DAYS OR ORDER WILL BE CANCELLED
Rabies Vaccine, Killed Virus, dogs and cats 3 months and older. DOI is 1 year. Preservative: Gentamicin.