Learn More
Medchemexpress LLC HY-17359 25mg , Deferasirox CAS:201530-41-8 Purity:>98%
SDP
Catalog No. 502260999
Encompass_Preferred
HY-17359 25mg Deferasirox CAS: 201530-41-8 Deferasirox (ICL 670) is an orally available iron chelator used for the management of transfusional iron overload. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
This item is not returnable. View return policy
Catalog No. 50-226-0999 Supplier Medchemexpress LLC Supplier No. HY1735925MG
HY-17359 25mg Deferasirox CAS: 201530-41-8 Deferasirox (ICL 670) is an orally available iron chelator used for the management of transfusional iron overload. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.