Learn More
Cayman Chemical Deferasirox-d4, Purity: ≥99% deuterated forms (d1-d4), CAS: 1133425-75-8, 1 mg
SDP
Catalog No. 502936850
Encompass_Preferred
An internal standard for the quantification of deferasirox by GC- or LC-MS
Catalog No. 50-293-6850 Supplier Cayman Chemical Supplier No. 254301 MG
An internal standard for the quantification of deferasirox by GC- or LC-MS