Learn More
Medchemexpress LLC HY-16564 5mg Medchemexpress, Deferasirox (Fe3+ chelate) CAS:554435-83-5 Purity:>98%
SDP
Catalog No. 502081893
Encompass_Preferred
Medchemexpress, HY-16564 5mg Deferasirox (Fe3+ chelate) CAS:554435-83-5 Deferasirox Fe3+ chelate is a rationally-designed oral iron chelator; its main use is to reduce chronic iron overload in patients who are receiving long-term blood transfusions for conditions such as beta-thalassemia and other chronic anemias. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-1893 Supplier Medchemexpress LLC Supplier No. HY165645MG
Medchemexpress, HY-16564 5mg Deferasirox (Fe3+ chelate) CAS:554435-83-5 Deferasirox Fe3+ chelate is a rationally-designed oral iron chelator; its main use is to reduce chronic iron overload in patients who are receiving long-term blood transfusions for conditions such as beta-thalassemia and other chronic anemias. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.