Learn More
Apexbio Technology LLC DEFERASIROX-FE3-CHELATE-2MG
SDP
Catalog No. 5000568010
Encompass_Preferred
Deferasirox Fe3 chelate (CAS No 554435-83-5) is a critical compound in biomedical research primarily used in the study of iron overload disorders Originating from studies on iron chelation therapy its mechanism of action involves binding free iron ions to form a stable complex thereby preventing iron-mediated oxidative stress This chelation process targets pathways associated with iron homeostasis In vitro Deferasirox Fe3 chelate demonstrates activity in the nanomolar to low micromolar range indicating potent binding efficiency It is commonly applied in research using various cell models and animal studies to assess its efficacy and safety in chelation therapy contexts The compound is also utilized in drug screening efforts to develop treatments for conditions like thalassemia and sickle cell disease Experimental concentrations typically depend on the specific research design allowing flexibility in application across diverse experimental settings
Catalog No. 50-005-68010 Supplier Apexbio Technology LLC Supplier No. A33552
Deferasirox Fe3 chelate (CAS No 554435-83-5) is a critical compound in biomedical research primarily used in the study of iron overload disorders Originating from studies on iron chelation therapy its mechanism of action involves binding free iron ions to form a stable complex thereby preventing iron-mediated oxidative stress This chelation process targets pathways associated with iron homeostasis In vitro Deferasirox Fe3 chelate demonstrates activity in the nanomolar to low micromolar range indicating potent binding efficiency It is commonly applied in research using various cell models and animal studies to assess its efficacy and safety in chelation therapy contexts The compound is also utilized in drug screening efforts to develop treatments for conditions like thalassemia and sickle cell disease Experimental concentrations typically depend on the specific research design allowing flexibility in application across diverse experimental settings