Learn More
Medchemexpress LLC Deferasirox (Fe3+ chelate) | 554435-83-5 | 98.0% | 426.18 | 1 ML
SDP
Catalog No. 5000394185
Encompass_Preferred
Deferasirox Fe3+ Chelate is an iron chelating agent extracted from patent WO2003053986. It is for research use only and not sold to patients.
  • Iron chelating agent
  • High purity of 98.0%
  • Solid appearance ranging from light brown to black
  • Stable for 3 years at -20°C as powder, and 2 years at -80°C in solvent
  • Soluble in DMSO (50 mg/mL)
  • Applicable in disease areas such as infection and bacterial infection
  • Associated with apoptosis, anti-infection, and ferroptosis research
Catalog No. 50-003-94185 Supplier Medchemexpress LLC Supplier No. HY1656410MM/1ML
Deferasirox Fe3+ Chelate is an iron chelating agent extracted from patent WO2003053986. It is for research use only and not sold to patients.
  • Iron chelating agent
  • High purity of 98.0%
  • Solid appearance ranging from light brown to black
  • Stable for 3 years at -20°C as powder, and 2 years at -80°C in solvent
  • Soluble in DMSO (50 mg/mL)
  • Applicable in disease areas such as infection and bacterial infection
  • Associated with apoptosis, anti-infection, and ferroptosis research