Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Deferasirox (Fe3+ chelate) | 554435-83-5 | 98.0% | 426.18 | 100 MG
SDP

Catalog No. 5000394512
Encompass_Preferred

Deferasirox Fe3+ Chelate is an iron chelating agent primarily used for research purposes. It is extracted from patent WO2003053986.

  • Appearance: Solid
  • Color: Light brown to black
  • Solubility in DMSO: 50 mg/mL (117.32 mM)
  • Powder storage: -20°C for 3 years, 4°C for 2 years
  • In-solvent storage: -80°C for 2 years, -20°C for 1 year

Catalog No. 50-003-94512 Supplier Medchemexpress LLC Supplier No. HY16564100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Deferasirox Fe3+ Chelate is an iron chelating agent primarily used for research purposes. It is extracted from patent WO2003053986.

  • Appearance: Solid
  • Color: Light brown to black
  • Solubility in DMSO: 50 mg/mL (117.32 mM)
  • Powder storage: -20°C for 3 years, 4°C for 2 years
  • In-solvent storage: -80°C for 2 years, -20°C for 1 year

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.