Learn More
Medchemexpress LLC Deferasirox (Fe3+ chelate) | 554435-83-5 | 98.0% | 426.18 | 50 MG
SDP
Catalog No. 5000393834
Encompass_Preferred
Deferasirox Fe3+ Chelate is an iron chelating agent. It is intended for research use only, extracted from patent WO2003053986.
  • Iron chelating agent
  • Intended for research use only
  • Solid appearance with color ranging from light brown to black
  • Soluble in DMSO: 50 mg/mL (117.32 mM)
  • Store powder at -20°C for 3 years, 4°C for 2 years
  • Store in solvent at -80°C for 2 years, -20°C for 1 year
Catalog No. 50-003-93834 Supplier Medchemexpress LLC Supplier No. HY1656450MG
Deferasirox Fe3+ Chelate is an iron chelating agent. It is intended for research use only, extracted from patent WO2003053986.
  • Iron chelating agent
  • Intended for research use only
  • Solid appearance with color ranging from light brown to black
  • Soluble in DMSO: 50 mg/mL (117.32 mM)
  • Store powder at -20°C for 3 years, 4°C for 2 years
  • Store in solvent at -80°C for 2 years, -20°C for 1 year