Learn More
Cayman Chemical Deferiprone-d3, Purity: ≥99% deuterated forms (d1-d3), CAS: 1346601-82-8, 1 mg
SDP
Catalog No. 502941510
Encompass_Preferred
An internal standard for the quantification of deferiprone by GC- or LC-MS
Catalog No. 50-294-1510 Supplier Cayman Chemical Supplier No. 287021 MG
An internal standard for the quantification of deferiprone by GC- or LC-MS