Learn More
Medchemexpress LLC Deferiprone | 30652-11-0 | 99.98% | 500 MG
SDP
Catalog No. 5000407911
Encompass_Preferred
Deferiprone is a potent, orally active, brain-penetrant, cell-penetrant, skin-permeable, free iron chelating agent. It inhibits the proliferation and migration and stimulates apoptosis in tumor cells.
  • Exhibits antianemic, neuroprotective, anti-inflammatory, antioxidant, and antidotal activities.
  • Suitable for studies in cancer, cardiovascular disease, infection, inflammation, and neurological diseases.
Catalog No. 50-004-07911 Supplier Medchemexpress LLC Supplier No. HYB0568500MG
Deferiprone is a potent, orally active, brain-penetrant, cell-penetrant, skin-permeable, free iron chelating agent. It inhibits the proliferation and migration and stimulates apoptosis in tumor cells.
  • Exhibits antianemic, neuroprotective, anti-inflammatory, antioxidant, and antidotal activities.
  • Suitable for studies in cancer, cardiovascular disease, infection, inflammation, and neurological diseases.