Learn More
Selleck Chemical LLC Deferiprone 50mg 30652-11-0 CP20
SDP
Supplier:  Selleck Chemical LLC S406750MG
Encompass_Preferred
Deferiprone (CP20) is a chelating agent with an affinity for ferric ion (iron III),binds with ferric ions to form neutral 3:1 (deferiprone:iron) complexes that are stable over a wide range of pH values. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-136-3627