Learn More
Cayman Chemical Deferitazole, Purity: ≥95%, CAS: 945635-15-4, 5 mg
SDP
Catalog No. 502947800
Encompass_Preferred
An iron chelator; decreases the levels of iron in iron-citrate complexes at 50 µM; reduces the number of P. falciparum oocytes, an effect that can be prevented by the addition of ferric chloride, and the percent of flagellated P. falciparum male gametocytes in vitro at 25 µM; decreases the urine and fecal levels of iron excreted from iron-loaded C. apella monkeys at 150 µmol/kg
Catalog No. 50-294-7800 Supplier Cayman Chemical Supplier No. 333295 MG
An iron chelator; decreases the levels of iron in iron-citrate complexes at 50 µM; reduces the number of P. falciparum oocytes, an effect that can be prevented by the addition of ferric chloride, and the percent of flagellated P. falciparum male gametocytes in vitro at 25 µM; decreases the urine and fecal levels of iron excreted from iron-loaded C. apella monkeys at 150 µmol/kg