Learn More
Medchemexpress LLC Deferoxamine | 70-51-9 | 99.6% | 100 MG
SDP
Catalog No. 5000413229
Encompass_Preferred
Deferoxamine is an iron chelator that binds to Fe(III) and other metal cations, used to reduce iron accumulation and deposition in tissues. It upregulates HIF-1α levels and exhibits good antioxidant activity. This compound also shows anti-proliferative activity and can induce apoptosis and autophagy in cancer cells.
  • Used in studies of diabetes.
  • Used in studies of neurodegenerative diseases.
  • Used in anti-cancer research.
  • Used in anti-COVID-19 research.
Catalog No. 50-004-13229 Supplier Medchemexpress LLC Supplier No. HYB1625100MG
Deferoxamine is an iron chelator that binds to Fe(III) and other metal cations, used to reduce iron accumulation and deposition in tissues. It upregulates HIF-1α levels and exhibits good antioxidant activity. This compound also shows anti-proliferative activity and can induce apoptosis and autophagy in cancer cells.
  • Used in studies of diabetes.
  • Used in studies of neurodegenerative diseases.
  • Used in anti-cancer research.
  • Used in anti-COVID-19 research.