Learn More
Medchemexpress LLC HY-B0988 500mg , Deferoxamine (mesylate) Desferrioxamine B mesylate;DFOM CAS:138-14-7 Purity:98%
SDP
Supplier:  Medchemexpress LLC HYB0988500MG
Encompass_Preferred
Medchemexpress, HY-B0988 500mg Deferoxamine (mesylate) Desferrioxamine B mesylate;DFOM CAS:138-14-7 Formula:C26H52N6O11S Purity:98% Deferoxamine mesylate is an iron chelator, and effectively reduces oxidative stress and neuronal death. Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-149-2367
 Restricted Item