Learn More
Medchemexpress LLC Deferoxamine (mesylate) | 138-14-7 | 99.8% | 20 MG
SDP
Catalog No. 5000407536
Encompass_Preferred
Deferoxamine mesylate is an iron chelator that binds to Fe(III) and other metal cations. It is widely used to reduce iron accumulation and deposition in tissues. This compound upregulates HIF-1α levels with good antioxidant activity, shows anti-proliferative activity, and can induce apoptosis and autophagy in cancer cells. It is applicable in studies of diabetes, neurodegenerative diseases, anti-cancer research, and anti-COVID-19 research.
  • Binds to Fe(III) and other metal cations
  • Reduces iron accumulation and deposition in tissues
  • Upregulates HIF-1α levels
  • Exhibits good antioxidant activity
  • Shows anti-proliferative activity
  • Induces apoptosis and autophagy in cancer cells
  • Applicable in studies related to diabetes, neurodegenerative diseases, anti-cancer research, and anti-COVID-19 research
Catalog No. 50-004-07536 Supplier Medchemexpress LLC Supplier No. HYB098820MG
Deferoxamine mesylate is an iron chelator that binds to Fe(III) and other metal cations. It is widely used to reduce iron accumulation and deposition in tissues. This compound upregulates HIF-1α levels with good antioxidant activity, shows anti-proliferative activity, and can induce apoptosis and autophagy in cancer cells. It is applicable in studies of diabetes, neurodegenerative diseases, anti-cancer research, and anti-COVID-19 research.
  • Binds to Fe(III) and other metal cations
  • Reduces iron accumulation and deposition in tissues
  • Upregulates HIF-1α levels
  • Exhibits good antioxidant activity
  • Shows anti-proliferative activity
  • Induces apoptosis and autophagy in cancer cells
  • Applicable in studies related to diabetes, neurodegenerative diseases, anti-cancer research, and anti-COVID-19 research