Learn More
Medchemexpress LLC Deferoxamine mesylate (standard) | 138-14-7 | 99.47% | 50 MG
SDP
Catalog No. 5000407537
Encompass_Preferred
Deferoxamine mesylate (Standard) is an analytical standard intended for research and analytical applications. It functions as an iron chelator, binding to Fe(III) and other metal cations, thereby reducing iron accumulation and deposition in tissues. This compound also upregulates HIF-1α levels and exhibits good antioxidant activity, along with anti-proliferative activity that can induce apoptosis and autophagy in cancer cells.
  • Used in studies related to diabetes, neurodegenerative diseases, anti-cancer research, and anti-COVID-19 research
  • Serves as a reference standard for assays
  • Frequently utilized in qualitative, quantitative, and methodological research experiments such as HPLC, GC, and MS
Catalog No. 50-004-07537 Supplier Medchemexpress LLC Supplier No. HYB0988R50MG
Deferoxamine mesylate (Standard) is an analytical standard intended for research and analytical applications. It functions as an iron chelator, binding to Fe(III) and other metal cations, thereby reducing iron accumulation and deposition in tissues. This compound also upregulates HIF-1α levels and exhibits good antioxidant activity, along with anti-proliferative activity that can induce apoptosis and autophagy in cancer cells.
  • Used in studies related to diabetes, neurodegenerative diseases, anti-cancer research, and anti-COVID-19 research
  • Serves as a reference standard for assays
  • Frequently utilized in qualitative, quantitative, and methodological research experiments such as HPLC, GC, and MS