Learn More
Cayman Chemical DeferoxamInmesylate 1g
SDP
Catalog No. 502968516
Encompass_Preferred
A bacterial siderophore that chelates iron; used experimentally to inhibit iron-dependent prolyl hydroxylases (EC50 = 17.8 µM), thus preventing the degradation of isoforms of HIF during normoxia
Catalog No. 50-296-8516 Supplier Cayman Chemical Supplier No. 145951 G
A bacterial siderophore that chelates iron; used experimentally to inhibit iron-dependent prolyl hydroxylases (EC50 = 17.8 µM), thus preventing the degradation of isoforms of HIF during normoxia