Learn More
Alkali Scientific Defibrinated Sheep Blood 100mL, Glass 4oz Crimp Bottle
SDP
Catalog No. 5000663110
Encompass_Preferred
Defibrinated Sheep Blood 100mL Glass 4oz Crimp Bottle is a sterile blood product used as a culture media additive to support the growth of fastidious microorganisms and to enable hemolysis testing in microbiological laboratories. The defibrination process removes fibrin to prevent clotting, allowing smooth and uniform mixing when incorporated into agar or broth media formulations. Sheep blood is widely used for preparing enriched media such as blood agar and chocolate agar for clinical, research, and diagnostic microbiology applications. Each bottle contains 100mL of sterile defibrinated sheep blood packaged in a glass crimp sealed container to maintain sterility, product stability, and convenient handling during routine laboratory media preparation.
Catalog No. 50-006-63110 Supplier Alkali Scientific Supplier No. DSBB00
Defibrinated Sheep Blood 100mL Glass 4oz Crimp Bottle is a sterile blood product used as a culture media additive to support the growth of fastidious microorganisms and to enable hemolysis testing in microbiological laboratories. The defibrination process removes fibrin to prevent clotting, allowing smooth and uniform mixing when incorporated into agar or broth media formulations. Sheep blood is widely used for preparing enriched media such as blood agar and chocolate agar for clinical, research, and diagnostic microbiology applications. Each bottle contains 100mL of sterile defibrinated sheep blood packaged in a glass crimp sealed container to maintain sterility, product stability, and convenient handling during routine laboratory media preparation.