Learn More
Thermo Scientific™ Defibrinated Sheep Blood
Description
Be confident that your animal blood meets the highest standards possible by using Thermo Scientific™ Defibrinated Sheep Blood, sourced from our own technologically-advanced sheep farm.
Specifications
Specifications
| Species | Sheep |
| Form | Liquid |
| Product Type | Whole Blood |
| Quantity | 250 mL |
| Processing | Defibrinated |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.