Learn More
FUJIFILM Biosciences Deficient DME High Glucose
Catalog No. 5000450259
Encompass
Dulbecco s Modified Eagle s Medium DMEM is a modification of Basal Medium Eagle BME containing two to fourfold higher concentrations of glucose amino acids and vitamins to better support highly proliferative cells such as embryonic cells
Catalog No. 50-004-50259 Supplier FUJIFILM Biosciences Supplier No. 9041500ML
Dulbecco s Modified Eagle s Medium DMEM is a modification of Basal Medium Eagle BME containing two to fourfold higher concentrations of glucose amino acids and vitamins to better support highly proliferative cells such as embryonic cells