Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Description
- Definitive endoderm induction medium is a defined, animal component-free, ready-to-use serum-free medium for efficient differentiation of human pluripotent stem cells (ES/iPSCs) to definitive endoderm (DE) cells
- No growth
Specifications
Specifications
| Form | Liquid |
| For Use With (Application) | Stem Cell Culture, Cell Culture |
| Quantity | 50 mL |
| Assays | <85% CXCR4 + c-Kit + double positive after 3 days of induction |
| Endotoxin Level | <2 EU/mL |
| pH | 7.0 to 7.4 |
| Content And Storage | Upon receipt -20 °C, Ready to use 2 to 8 ° overnight thaw, Use immediately store (at 2°C to 8°C up to 1 week, Do not re-freeze) |
| Description | Clear liquid/no particulates |
For research purposes only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?