Learn More
Cayman Chemical Deflazacort, Purity: ≥98%, CAS: 14484-47-0, 100 mg
SDP
Catalog No. 502982503
Encompass_Preferred
A prodrug for the active glucocorticoid 21-desacetyl deflazacort
Catalog No. 50-298-2503 Supplier Cayman Chemical Supplier No. 20386100 MG
A prodrug for the active glucocorticoid 21-desacetyl deflazacort