Learn More
Medchemexpress LLC Deflazacort-d3
SDP
Catalog No. 5000284806
Encompass_Preferred
Deflazacort-d3 is a isotope of Deflazacort Deflazacort is an inactive proagent and is converted rapidly to the active metabolite 21-desacetyldeflazacort Deflazacort is used as an anti-inflammatory and immunosuppressant[1]
Catalog No. 50-002-84806 Supplier Medchemexpress LLC Supplier No. HYW6543975MG
Deflazacort-d3 is a isotope of Deflazacort Deflazacort is an inactive proagent and is converted rapidly to the active metabolite 21-desacetyldeflazacort Deflazacort is used as an anti-inflammatory and immunosuppressant[1]