Learn More
Enterprise Technology Solutions Deflecto Mounting Bar, Storage Organizer, 22" L, Gray
SDP
Catalog No. 5000831493
Encompass_Preferred
Wall-Mounting Bar features an easy-to-use design to make installation fast and simple. Durable aluminum construction is capable of holding up to 40 lb. to keep your supplies in a convenient location. This 22" mounting bar is designed for use with Deflecto Stackable Caddy Organizer Canisters and Interlocking Single Tilt Bins (each not included) to store a variety of essential items in one place.
Catalog No. 50-008-31493 Supplier Enterprise Technology Solutions Supplier No. DEF22773CR
Wall-Mounting Bar features an easy-to-use design to make installation fast and simple. Durable aluminum construction is capable of holding up to 40 lb. to keep your supplies in a convenient location. This 22" mounting bar is designed for use with Deflecto Stackable Caddy Organizer Canisters and Interlocking Single Tilt Bins (each not included) to store a variety of essential items in one place.