Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Deflouro-(p-tolylsulfonyloxy)-N-boc-florbetapir | 1205550-99-7 | 99.7% | 50 MG
SDP

Catalog No. 5000232152
Encompass_Preferred

AV-105 is a tosylate precursor used to prepare 18F-radiolabeled florbetapir derivatives for positron emission tomography (PET) imaging research. It is provided as a white to yellow solid with high purity and is intended for use as a synthetic intermediate in radiochemistry workflows.

  • High purity: 99.7%.
  • Molecular weight: 612.7 g/mol.
  • Chemical formula: C32H40N2O8S.
  • Appearance: white to yellow solid.
  • Intended use: synthetic intermediate for preparing 18F-radiolabeled PET imaging agents (research use only).
  • Storage: powder -20°C (3 years) or 4°C (2 years); in solvent -80°C (2 years) or -20°C (1 year).

Catalog No. 50-002-32152 Supplier Medchemexpress LLC Supplier No. HY12094750MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

AV-105 is a tosylate precursor used to prepare 18F-radiolabeled florbetapir derivatives for positron emission tomography (PET) imaging research. It is provided as a white to yellow solid with high purity and is intended for use as a synthetic intermediate in radiochemistry workflows.

  • High purity: 99.7%.
  • Molecular weight: 612.7 g/mol.
  • Chemical formula: C32H40N2O8S.
  • Appearance: white to yellow solid.
  • Intended use: synthetic intermediate for preparing 18F-radiolabeled PET imaging agents (research use only).
  • Storage: powder -20°C (3 years) or 4°C (2 years); in solvent -80°C (2 years) or -20°C (1 year).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.