Learn More
Chemworld Defoamer / Antifoam (Silicone Based) - 5 Gallons
SDP
Catalog No. NC1762115
Encompass
ChemWorld S, Silicone Defoamer, is a 10% active silicone antifoam for a wide range of applications. It is useful in process, waste treatment, cleaning and certain other applications where silicone can be used. This product is an excellent, fast acting material for surfactant-generated foams which can cause cleaning or treatment problems. In systems where continuous agitation and/or source of foam causing agents exist, continuous treatment will be required.
Catalog No. NC1762115 Supplier Chemworld Supplier No. DEFOAMERS5
ChemWorld S, Silicone Defoamer, is a 10% active silicone antifoam for a wide range of applications. It is useful in process, waste treatment, cleaning and certain other applications where silicone can be used. This product is an excellent, fast acting material for surfactant-generated foams which can cause cleaning or treatment problems. In systems where continuous agitation and/or source of foam causing agents exist, continuous treatment will be required.