Learn More
Apexbio Technology LLC DEGARELIX-ACETATE-25MG
SDP
Catalog No. 5000567542
Encompass_Preferred
Degarelix acetate (CAS No 934016-19-0) is a gonadotropin-releasing hormone (GnRH) antagonist used in biomedical research primarily for its role in androgen deprivation therapy Originating from synthetic peptides it functions by binding to GnRH receptors in the anterior pituitary gland effectively decreasing the release of luteinizing hormone and subsequently reducing testosterone production Its in vitro activity typically ranges from the nanomolar to low micromolar concentration Degarelix acetate is commonly applied in cell models and animal studies to investigate prostate cancer treatment and it is also used in drug screening for hormonal therapies Experimental concentrations typically depend on the specific research design offering flexibility in application across various studies
Catalog No. 50-005-67542 Supplier Apexbio Technology LLC Supplier No. C871825
Degarelix acetate (CAS No 934016-19-0) is a gonadotropin-releasing hormone (GnRH) antagonist used in biomedical research primarily for its role in androgen deprivation therapy Originating from synthetic peptides it functions by binding to GnRH receptors in the anterior pituitary gland effectively decreasing the release of luteinizing hormone and subsequently reducing testosterone production Its in vitro activity typically ranges from the nanomolar to low micromolar concentration Degarelix acetate is commonly applied in cell models and animal studies to investigate prostate cancer treatment and it is also used in drug screening for hormonal therapies Experimental concentrations typically depend on the specific research design offering flexibility in application across various studies