Learn More
Medchemexpress LLC HY-16168A 100mg , Degarelix CAS:214766-78-6 Purity:98%
SDP
Supplier:  Medchemexpress LLC HY16168A100MG
Encompass_Preferred
Medchemexpress, HY-16168A 100mg Degarelix CAS:214766-78-6 Formula:C82H103ClN18O16 Purity:98% Degarelix is a competitive gonadotropin-releasing hormone receptor ( GnRHR ) antagonist for the treatment of androgen-dependent advanced prostate cancer. Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-149-2896