Learn More
Advanced Biomatrix Degassed Water, 10 x 10 mL
Catalog No. 501960321
Encompass_Preferred
Sterile filtered, degassed, deionized water at pH 5.0-8.0. This product was designed and intended for use with the HyStem family of thiolated hyaluronic acid products.
Catalog No. 50-196-0321 Supplier Advanced Biomatrix Supplier No. GS250F10EA
Sterile filtered, degassed, deionized water at pH 5.0-8.0. This product was designed and intended for use with the HyStem family of thiolated hyaluronic acid products.