Learn More
Waters Corp Waters Corporation Tubing Assembly, Degasser Port B2 to SSV B, 1/16 in. OD, 0.040 in. ID
Catalog No. 50791890 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This product is a tubing assembly designed to connect the Degasser Port B2 to the Solvent Select Valve B. It is an essential component for maintaining the performance of various ACQUITY BSM systems.
  • Connects Degasser Port B2 to Solvent Select Valve B.
  • Features an outer diameter of 1/16 in.
  • Features an inner diameter of 0.040 in.
  • Compatible with ACQUITY I-Class BSM.
  • Compatible with ACQUITY UPLC Binary Solvent Manager.
  • Compatible with ACQUITY I-Class PLUS BSM.
  • Compatible with ACQUITY H-Class PLUS Binary Solvent Manager.
Catalog No. 50-791-890 Supplier Waters Corp Supplier No. 430001113
This product is a tubing assembly designed to connect the Degasser Port B2 to the Solvent Select Valve B. It is an essential component for maintaining the performance of various ACQUITY BSM systems.
  • Connects Degasser Port B2 to Solvent Select Valve B.
  • Features an outer diameter of 1/16 in.
  • Features an inner diameter of 0.040 in.
  • Compatible with ACQUITY I-Class BSM.
  • Compatible with ACQUITY UPLC Binary Solvent Manager.
  • Compatible with ACQUITY I-Class PLUS BSM.
  • Compatible with ACQUITY H-Class PLUS Binary Solvent Manager.