Learn More
Medchemexpress LLC HY-13264 10mg , Degrasyn CAS:856243-80-6 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1326410MG
Encompass_Preferred
HY-13264 10mg Degrasyn CAS: 856243-80-6 Degrasyn (WP1130) is a cell-permeable deubiquitinase (DUB) inhibitor, directly inhibiting DUB activity of USP9x, USP5, USP14, and UCH37. Degrasyn has been shown to downregulate the antiapoptotic proteins Bcr-Abl and JAK2. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-226-0731