Learn More
MSC Rite-Kem All-Purpose Cleaner & Degreaser, 55 gal, Plastic Drum, Dilutable Concentrate
Catalog No. 502837681
Encompass
This all-purpose cleaner and degreaser is a concentrated, water-dilutable liquid formula. Designed for multiple applications, it comes in a 55-gallon plastic drum and features a pleasant scent.
  • Concentrated degreasing formula
  • Water dilutable multi-purpose cleaner and degreaser
  • Liquid form
  • Biodegradable and water-based solution
  • Comes in a 55 gal plastic drum
  • Features a pleasant scent
  • Low VOC content of 5%
Catalog No. 50-283-7681 Supplier MSC Supplier No. 48175699
This all-purpose cleaner and degreaser is a concentrated, water-dilutable liquid formula. Designed for multiple applications, it comes in a 55-gallon plastic drum and features a pleasant scent.
  • Concentrated degreasing formula
  • Water dilutable multi-purpose cleaner and degreaser
  • Liquid form
  • Biodegradable and water-based solution
  • Comes in a 55 gal plastic drum
  • Features a pleasant scent
  • Low VOC content of 5%