Learn More
MSC Rite-Kem All-Purpose Cleaner, 32 oz Bottle, Liquid Concentrate, Pleasant Scent
Catalog No. 502839175
Encompass
The Rite-Kem All-Purpose Cleaner and Degreaser is a versatile liquid concentrate designed for effective cleaning. This water-based, biodegradable formula comes in a 32 oz bottle with a pleasant scent. It is non-acidic and contains 5% VOC, making it suitable for various cleaning tasks.
  • All-purpose cleaner and degreaser
  • Liquid concentrate form
  • 32 oz bottle
  • Pleasant scent
  • Not a disinfectant
  • Biodegradable formula
  • Water-based solution
  • Non-acidic
  • 5% VOC content
  • Made in the United States
Catalog No. 50-283-9175 Supplier MSC Supplier No. 48175640
The Rite-Kem All-Purpose Cleaner and Degreaser is a versatile liquid concentrate designed for effective cleaning. This water-based, biodegradable formula comes in a 32 oz bottle with a pleasant scent. It is non-acidic and contains 5% VOC, making it suitable for various cleaning tasks.
  • All-purpose cleaner and degreaser
  • Liquid concentrate form
  • 32 oz bottle
  • Pleasant scent
  • Not a disinfectant
  • Biodegradable formula
  • Water-based solution
  • Non-acidic
  • 5% VOC content
  • Made in the United States