Learn More
MSC Zep Electrical Cleaner & Degreaser, 22 oz, 13,800V, Non-Chlorinated
Catalog No. 502837283
Encompass
The Zep Selig Nu Tri Clean is a solvent-based cleaner and degreaser designed to effectively remove oils, greases, and adhesive film from various surfaces. It is a non-chlorinated, non-flammable, and fast-evaporating formula, making it suitable for sensitive applications such as aircraft parts, electrical components, and printed circuit boards. The cleaner leaves no residue and requires no rinsing.
  • Non-chlorinated and non-flammable formula.
  • Fast-evaporating with no residue, eliminating the need for rinsing.
  • Safe for use on aluminum, magnesium, and ferrous metals.
  • Effective for electrical parts cleaning and degreasing.
  • High dielectric strength of 13,800V.
Catalog No. 50-283-7283 Supplier MSC Supplier No. 65461212
The Zep Selig Nu Tri Clean is a solvent-based cleaner and degreaser designed to effectively remove oils, greases, and adhesive film from various surfaces. It is a non-chlorinated, non-flammable, and fast-evaporating formula, making it suitable for sensitive applications such as aircraft parts, electrical components, and printed circuit boards. The cleaner leaves no residue and requires no rinsing.
  • Non-chlorinated and non-flammable formula.
  • Fast-evaporating with no residue, eliminating the need for rinsing.
  • Safe for use on aluminum, magnesium, and ferrous metals.
  • Effective for electrical parts cleaning and degreasing.
  • High dielectric strength of 13,800V.