Learn More
Essendant DEGREASERGREZOFF12/32OZ
Catalog No. 5000032774
Encompass_Preferred
Quickly and easily degrease the gunkiest engines and parts Fast-acting surfactants penetrate permeate and remove grime heavy grease lubricants crude oil hydraulic fluid tar and more The degreaser formula is water-based solvent-free non-combustible non-flammable and fume-free Application- Degreaser/Cleaner Applicable Material- Asphalt Ceramics Cement/Concrete Conveyors Enamel Engines Fiberglass Grills Industrial and Farm Equipment Laminates Machinery Metal Mowers Ovens Plaster Plastic Rubber RVs Tools Trailers Trunks Vents Vinyl Dirt Types- Grease Grime Hydraulic Fluid Lubricants Oil-Based Soil Tar Scent- Unscented
Catalog No. 50-000-32774 Supplier Essendant Supplier No. ITW22732
Quickly and easily degrease the gunkiest engines and parts Fast-acting surfactants penetrate permeate and remove grime heavy grease lubricants crude oil hydraulic fluid tar and more The degreaser formula is water-based solvent-free non-combustible non-flammable and fume-free Application- Degreaser/Cleaner Applicable Material- Asphalt Ceramics Cement/Concrete Conveyors Enamel Engines Fiberglass Grills Industrial and Farm Equipment Laminates Machinery Metal Mowers Ovens Plaster Plastic Rubber RVs Tools Trailers Trunks Vents Vinyl Dirt Types- Grease Grime Hydraulic Fluid Lubricants Oil-Based Soil Tar Scent- Unscented