Learn More
MSC Rite-Kem All-Purpose Cleaner & Degreaser, Liquid Concentrate, Pleasant Scent, 4 gal
Catalog No. 502837680
Encompass
This all-purpose cleaner and degreaser is a concentrated, water-based, and biodegradable liquid solution. It features a pleasant scent and is designed for effective cleaning and degreasing applications.
  • Liquid concentrate
  • Pleasant scent
  • Biodegradable formula
  • Water-based solution
  • Dilutable for various cleaning tasks
  • Supplied in 1 gallon bottles
  • Non-caustic
  • VOC content of 5%
Catalog No. 50-283-7680 Supplier MSC Supplier No. 48175665
This all-purpose cleaner and degreaser is a concentrated, water-based, and biodegradable liquid solution. It features a pleasant scent and is designed for effective cleaning and degreasing applications.
  • Liquid concentrate
  • Pleasant scent
  • Biodegradable formula
  • Water-based solution
  • Dilutable for various cleaning tasks
  • Supplied in 1 gallon bottles
  • Non-caustic
  • VOC content of 5%