Learn More
MSC Rite-Kem All-Purpose Cleaner, 5 gal, Liquid Concentrate, Biodegradable
Catalog No. 502837227
Encompass
This all-purpose cleaner is a 5-gallon liquid concentrate designed for various cleaning and degreasing tasks. It is formulated to be biodegradable and water-based, offering effective cleaning without harsh acids or disinfectant properties. The dilutable concentrate features a pleasant scent and is suitable for use as a cleaning compound and solvent-detergent.
  • Liquid concentrate formula
  • Packaged in a 5 gal pail
  • Features a pleasant scent
  • Biodegradable and water-based
  • Non-acidic and non-disinfectant
  • Dilutable for various applications
  • Suitable for asphalt surfaces
Catalog No. 50-283-7227 Supplier MSC Supplier No. 48175798
This all-purpose cleaner is a 5-gallon liquid concentrate designed for various cleaning and degreasing tasks. It is formulated to be biodegradable and water-based, offering effective cleaning without harsh acids or disinfectant properties. The dilutable concentrate features a pleasant scent and is suitable for use as a cleaning compound and solvent-detergent.
  • Liquid concentrate formula
  • Packaged in a 5 gal pail
  • Features a pleasant scent
  • Biodegradable and water-based
  • Non-acidic and non-disinfectant
  • Dilutable for various applications
  • Suitable for asphalt surfaces