Learn More
Aviva Systems Biology Corp DEGS1 Antibody - N-terminal region (ARP45501_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP45501_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against DEGS1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human DEGS1. Alternate names: DEGS, DES1, Des-1, FADS7, MGC5079, MIG15, MLD, DEGS-1ApplicationWB, IHC. Species reactivity: Cow, Goat, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Sheep, Zebrafish
Catalog No. 50-014-59044