Learn More
Medchemexpress LLC Deguelin | 522-17-8 | MFCD01740600 | 98.0% | C23H22O6 | 10MG
SDP
Catalog No. 5000261387
Encompass_Preferred
Deguelin is a naturally occurring rotenoid used as a research reagent with reported activity against PI3K/Akt signaling and mitochondrial complex I. It is supplied as a solid analytical standard suitable for biochemical and cell biology studies.
  • Naturally occurring rotenoid with bioactivity against PI3K/Akt and mitochondrial complex I.
  • Suitable as an analytical standard for biochemical and cell biology assays.
  • High purity (~98%) for research applications.
  • Solid powder form, convenient for small-scale experiments.
  • Molecular formula C23H22O6 and molecular weight 394.42 g/mol.
Catalog No. 50-002-61387 Supplier Medchemexpress LLC Supplier No. HY1342510MG
Deguelin is a naturally occurring rotenoid used as a research reagent with reported activity against PI3K/Akt signaling and mitochondrial complex I. It is supplied as a solid analytical standard suitable for biochemical and cell biology studies.
  • Naturally occurring rotenoid with bioactivity against PI3K/Akt and mitochondrial complex I.
  • Suitable as an analytical standard for biochemical and cell biology assays.
  • High purity (~98%) for research applications.
  • Solid powder form, convenient for small-scale experiments.
  • Molecular formula C23H22O6 and molecular weight 394.42 g/mol.