Learn More
Cayman Chemical (-)-Deguelin, Purity: ≥98%, CAS: 522-17-8, 25 mg
SDP
Catalog No. 502890338
Encompass_Preferred
A rotenoid with diverse biological activities; inhibits mitochondrial complex I activity in bovine heart electron transport particles and phorbol 12-myristate 13-acetate-induced ODC activity in MCF-7 cells (IC50s = 6.9 and 11 nM, respectively); induces apoptosis in MCF-7 and SK-BR-3 cells at 100 nM; induces mortality in adult bruchid beetles (C. maculatus) from 10-500 ppm; induces nigrostriatal dopaminergic degeneration in rats at 6 mg/kg
Catalog No. 50-289-0338 Supplier Cayman Chemical Supplier No. 1001070625 MG
A rotenoid with diverse biological activities; inhibits mitochondrial complex I activity in bovine heart electron transport particles and phorbol 12-myristate 13-acetate-induced ODC activity in MCF-7 cells (IC50s = 6.9 and 11 nM, respectively); induces apoptosis in MCF-7 and SK-BR-3 cells at 100 nM; induces mortality in adult bruchid beetles (C. maculatus) from 10-500 ppm; induces nigrostriatal dopaminergic degeneration in rats at 6 mg/kg