Learn More
CHEMetrics™ by AquaPhoenix DEHA/Carbohydrazide Refill Kit

Description
- Sample is treated with an excess of ferric iron.
- DEHA (N,N-diethylhydroxylamine) reacts quantitatively with ferric iron by reducing it to the ferrous state. The resulting ferrous iron reacts with PDTS [3-(2-pyridyl)-5,6-bis(4-phenylsulfonic acid)-1,2,4,- triazine disodium salt] to form a pink-purple colored complex in direct proportion to the DEHA concentration. Test results are expressed as ppb (μg/L) or ppm (mg/L) DEHA.
Specifications
Specifications
| pH Range | 7 |
| Volume (Metric) | 0.5 mL |
| For Use With (Equipment) | K-3902 or K-1805 |
| No. of Tests | 30 |
| Quantity | 30/Box |
| Type | DEHA/Carbohydrazide Refill Kit |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.