Learn More
Hach Company DEHA Reagent 1 Powder Pillows, pk/100
Supplier:  Hach Company 2167969
Encompass
For determination of oxygen scavengers by the iron reduction method. Hach method 8140 determines carbohydrazide, diethylhydroxylamine (DEHA), hydroquinone, iso-ascorbic acid (erythorbic acid) and methylethylketoxime. Pack of 100 Powder Pillows.
Catalog No. 50-199-5497