Learn More
Hach Company DEHA Reagent 2 Solution, 500 mL
Supplier:  Hach Company 2168049
Encompass
For determination of oxygen scavengers by the iron reduction method. Hach method 8140 determines carbohydrazide, diethylhydroxylamine (DEHA), hydroquinone, iso-ascorbic acid (erythorbic acid) and methylethylketoxime. 500 mL
Catalog No. 50-199-5499