Learn More
CHEMetrics™ by AquaPhoenix DEHA Water Quality Visual Test Kit
SDP
Catalog No. 13299560
Change view
Click to view available options
No. of Tests:
30
Catalog No. No. of Tests
13-299-560 30
1 options
Catalog No. 13-299-560 Supplier CHEMetrics™ by AquaPhoenix Supplier No. K3902
Add to Cart
Add to Cart
DEHA visual test kit employing PDTS chemistry for colorimetric analysis. Measures DEHA in the 0-400 ppb and 400-3000 ppb ranges.
  • The kit comes in a plastic case and contains everything needed to perform 30 tests
  • Refill, low and high range comparators, activator solution, 25 ml sample cup, and instructions.
Specifications
Analytes DEHA
pH Range 7
Volume (Metric) 0.5 mL
No. of Tests 30
Range 0 to 400 and 400 to 3000 ppb
Test Method PDTS
Type Water Quality Visual Test Kit