Learn More
ATCC Acinetobacter baumannii; Strain A118
Catalog No. 502381256
Encompass
Genome sequenced strain. Possesses the blaOXA-89 gene.
Catalog No. 50-238-1256 Supplier ATCC Supplier No. BAA2093
Genome sequenced strain. Possesses the blaOXA-89 gene.