Learn More
Medchemexpress LLC DEHP-d4 (Bis(2-ethylhexyl) phthalate-d4) | 93951-87-2 | 99.9% | 1 MG
SDP
Catalog No. 5000412190
Encompass_Preferred
DEHP-d4 is the deuterium labeled version of DEHP, an endogenous metabolite. It is primarily used as a tracer and an internal standard for quantitative analysis. This compound is for research use only.
  • Used as a tracer
  • Functions as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Incorporates stable heavy isotopes of hydrogen and carbon
  • Deuteration can affect the pharmacokinetic and metabolic profiles of drugs
Catalog No. 50-004-12190 Supplier Medchemexpress LLC Supplier No. HYB1945S1MG
DEHP-d4 is the deuterium labeled version of DEHP, an endogenous metabolite. It is primarily used as a tracer and an internal standard for quantitative analysis. This compound is for research use only.
  • Used as a tracer
  • Functions as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Incorporates stable heavy isotopes of hydrogen and carbon
  • Deuteration can affect the pharmacokinetic and metabolic profiles of drugs