Learn More
Medchemexpress LLC DEHP-d4 (bis(2-ethylhexyl) phthalate-d4) | 93951-87-2 | 99.9% | 25 MG
SDP
Catalog No. 5000411254
Encompass_Preferred
DEHP-d4 is the deuterium labeled DEHP, an endogenous metabolite. This compound is intended for research use only and can be utilized as a tracer or an internal standard for quantitative analysis. It is suitable for analytical techniques such as NMR, GC-MS, or LC-MS.
  • Used as a tracer
  • Used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Stable heavy isotopes incorporated into drug molecules for quantitation
  • Potential to affect pharmacokinetic and metabolic profiles of drugs
Catalog No. 50-004-11254 Supplier Medchemexpress LLC Supplier No. 25MGHYB1945S
DEHP-d4 is the deuterium labeled DEHP, an endogenous metabolite. This compound is intended for research use only and can be utilized as a tracer or an internal standard for quantitative analysis. It is suitable for analytical techniques such as NMR, GC-MS, or LC-MS.
  • Used as a tracer
  • Used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Stable heavy isotopes incorporated into drug molecules for quantitation
  • Potential to affect pharmacokinetic and metabolic profiles of drugs