Learn More
Medchemexpress LLC DEHP-d4 (Bis(2-ethylhexyl) phthalate-d4) | 93951-87-2 | MFCD00143759 | 99.9% | 1 ML
SDP
Catalog No. 5000412169
Encompass_Preferred
DEHP-d4 is a deuterium-labeled variant of Bis(2-ethylhexyl) phthalate (DEHP), an endogenous metabolite. This compound is intended for research purposes, specifically as a tracer or an internal standard for quantitative analysis through methods such as NMR, GC-MS, or LC-MS. Deuterium substitution in drug molecules has been noted for its potential to alter pharmacokinetic and metabolic profiles.
Features and Benefits:
  • Used as a tracer.
  • Serves as an internal standard for quantitative analysis.
  • Applicable in NMR, GC-MS, or LC-MS analysis.
  • Deuterium labeling can impact pharmacokinetic and metabolic profiles.
  • High purity of 99.9%.
Catalog No. 50-004-12169 Supplier Medchemexpress LLC Supplier No. HYB1945S10MM/1ML
DEHP-d4 is a deuterium-labeled variant of Bis(2-ethylhexyl) phthalate (DEHP), an endogenous metabolite. This compound is intended for research purposes, specifically as a tracer or an internal standard for quantitative analysis through methods such as NMR, GC-MS, or LC-MS. Deuterium substitution in drug molecules has been noted for its potential to alter pharmacokinetic and metabolic profiles.
Features and Benefits:
  • Used as a tracer.
  • Serves as an internal standard for quantitative analysis.
  • Applicable in NMR, GC-MS, or LC-MS analysis.
  • Deuterium labeling can impact pharmacokinetic and metabolic profiles.
  • High purity of 99.9%.