Learn More
Grainger Dehumidistat 10 A Silver 115 V
Catalog No. 502750545
Encompass_Preferred
The Tjernlund Dehumidistat Wall Mount is a wall-mounted humidistat that controls the operation of dehumidifiers, ventilation fans, and other humidity-controlled devices. It has an adjustable humidity setpoint from 30% to 100% RH. The dehumidistat features a rugged plastic enclosure and measures 4.5 x 2.75 x 1.25 inches. It requires a 24VAC power source and can switch up to 5 amps at 24VAC. The dehumidistat includes a built-in differential of 5% RH to prevent short cycling. It comes with a 6 foot power cord and has a limited 1 year warranty from Tjernlund.
Catalog No. 50-275-0545 Supplier Grainger Supplier No. 14U174
The Tjernlund Dehumidistat Wall Mount is a wall-mounted humidistat that controls the operation of dehumidifiers, ventilation fans, and other humidity-controlled devices. It has an adjustable humidity setpoint from 30% to 100% RH. The dehumidistat features a rugged plastic enclosure and measures 4.5 x 2.75 x 1.25 inches. It requires a 24VAC power source and can switch up to 5 amps at 24VAC. The dehumidistat includes a built-in differential of 5% RH to prevent short cycling. It comes with a 6 foot power cord and has a limited 1 year warranty from Tjernlund.