Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals™ Dehybernation Solution A Bulk

Catalog No. MP111002201 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
110 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP111002201 110 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MP111002201 Supplier MP Biomedicals™ Supplier No. 111002201
Add to Cart
Add to Cart

Dehybernation Solution A Bulk is a guanidine-based solution for the purification of DNA from ancient or aged samples such as bone and preserved tissue.

  • Research Use Only (RUO).
  • Ready for CE IVD labeling of clinical applications.
TRUSTED_SUSTAINABILITY

Specifications

CAS 593-84-0
Content And Storage AllgENECLEAN™ Kits are shipped at room temperature (15°C to 30°C).
For Use With (Application) DNA & RNA Purification
No. of Reactions 300 Preps
Packaging Type Bulk Packaged
pH 4.7 to 5.3
Quantity 110 mL
Synonym Rapid purification of linear DNA, DNA Purification Sample Preparation Solution
Form Liquid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.